Merck
  • Kinetic investigation of the OH radical and Cl atom initiated degradation of unsaturated ketones at atmospheric pressure and 298 K.
Kinetic investigation of the OH radical and Cl atom initiated degradation of unsaturated ketones at atmospheric pressure and 298 K.
The journal of physical chemistry. A (2012-01-26)
María B Blanco, Ian Barnes, Peter Wiesen
ABSTRACT
Rate coefficients for the reactions of hydroxyl radicals and chlorine atoms with 4-hexen-3-one, 5-hexen-2-one, and 3-penten-2-one have been determined at 298 ± 2 K and atmospheric pressure of air. Rate coefficients for the compounds were determined using a relative kinetic technique with different reference compounds. The experiments were performed in a large photoreactor (480 L) using in situ FTIR spectroscopy to monitor the decay of reactants. From the different measurements the following rate coefficients (in units of cm(3) molecule(-1) s(-1)) have been determined: k(1)(OH + 4-hexen-3-one) = (9.04 ± 2.12) × 10(-11), k(2)(OH + 5-hexen-2-one) = (5.18 ± 1.27) × 10(-11), k(3)(OH + 3-penten-2-one) = (7.22 ± 1.74) × 10(-11), k(4)(Cl + 4-hexen-3-one) = (3.00 ± 0.58) × 10(-10), k(5)(Cl + 5-hexen-2-one) = (3.15 ± 0.50) × 10(-10) and k(6)(Cl + 3-penten-2-one) = (2.53 ± 0.54) × 10(-10). The reactivity of the double bond in alkenes and unsaturated ketones at 298 K toward addition of OH radicals and Cl atoms are compared and discussed. In addition, a correlation between the reactivity of the unsaturated ketones toward OH radicals and the HOMO of the compounds is presented. On the basis of the kinetic measurements, the tropospheric lifetimes of 4-hexen-3-one, 5-hexen-2-one, and 3-penten-2-one with respect to their reaction with hydroxyl radicals are estimated to be between 2 and 3 h.