Merck
  • Immunophenotyping of inflammatory cells associated with Schmallenberg virus infection of the central nervous system of ruminants.
Immunophenotyping of inflammatory cells associated with Schmallenberg virus infection of the central nervous system of ruminants.
PloS one (2013-05-15)
Vanessa Herder, Florian Hansmann, Peter Wohlsein, Martin Peters, Mariana Varela, Massimo Palmarini, Wolfgang Baumgärtner
ABSTRACT
Schmallenberg virus (SBV) is a recently discovered Bunyavirus associated mainly with abortions, stillbirths and malformations of the skeletal and central nervous system (CNS) in newborn ruminants. In this study, a detailed immunophenotyping of the inflammatory cells of the CNS of affected animals was carried out in order to increase our understanding of SBV pathogenesis. A total of 82 SBV-polymerase chain reaction (PCR) positive neonatal ruminants (46 sheep lambs, 34 calves and 2 goat kids) were investigated for the presence of inflammation in the brain and spinal cord. The study focused on 15 out of 82 animals (18.3%) showing inflammation in the CNS. All 15 neonates displayed lymphohistiocytic meningoencephalomyelitis affecting most frequently the mesencephalon and the parietal and temporal lobes. The majority of infiltrating cells were CD3-positive T cells, followed by CD79α-positive B cells and CD68-positive microglia/macrophages. Malformations like por- and hydranencephaly, frequently found in the temporal lobe, showed associated demyelination and axonal loss. SBV antigen was detected in 37 out of 82 (45.1%) neonatal brains by immunohistochemistry. In particular, SBV antigen was found in 93.3% (14 out of 15 ruminants) and 32.8% (22 out of 67 ruminants) of animals with and without encephalitis, respectively. Highest amounts of virus-protein expression levels were found in the temporal lobe. Our findings suggest that: (i) different brain regions display differential susceptibility to SBV infection; (ii) inflammatory cells in the CNS are found only in a minority of virus infected animals; (iii) malformations occur in association with and without inflammation in the CNS; and (iv) viral antigen is strongly associated with the presence of inflammation in naturally infected animals. Further studies are required to explore the cell tropism and pathogenesis of SBV infection in ruminants.