Merck
Z103403
Swing-A Way portable can opener
heavy-duty drive gears and an extra-hard cutting wheel
Sign In to View Organizational & Contract Pricing
About This Item
UNSPSC Code:
41100000
Skip To
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
General description
Useful for opening the overpack cans in which hazardous or odoriferous compounds are shipped. Chrome-plated. Has heavy-duty drive gears and an extra-hard cutting wheel mounted on a wide-diameter permanent-alignment shaft.
Choose from one of the most recent versions:
Certificates of Analysis (COA)