Skip to Content

BDPH9BCD90FB

Bide Pharmatech

Methyl 2-(4-aminophenoxy)acetate

Synonym(s):

methyl (4-aminophenoxy)acetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H11O3N1
CAS Number:
Molecular Weight:
181.19
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H11NO3/c1-12-9(11)6-13-8-4-2-7(10)3-5-8/h2-5H,6,10H2,1H3

Inchi Key

YDWPOGYTJVQQIL-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid or semi-solid or liquid or lump



Still not finding the right product?

Explore all of our products under Methyl 2-(4-aminophenoxy)acetate


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library