Skip to Content
Merck

BDPH9BCDC8FC

Bide Pharmatech

1-(Quinoxalin-6-yl)ethanol

Synonym(s):

1-(6-quinoxalinyl)ethanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H10O1N2
CAS Number:
Molecular Weight:
174.2
MDL number:
Product BDPH9BCDC8FC is not currently sold in your country.


Inchi Code

1S/C10H10N2O/c1-7(13)8-2-3-9-10(6-8)12-5-4-11-9/h2-7,13H,1H3

Inchi Key

AZJBZDWQJDRHNH-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?

Explore all of our products under 1-(Quinoxalin-6-yl)ethanol



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library