Skip to Content

BDPH9C01FA2A

Bide Pharmatech

3-Bromocyclobutane-1-carboxylic acid

Synonym(s):

3-bromocyclobutanecarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H7O2Br1
CAS Number:
Molecular Weight:
179.01
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H7BrO2/c6-4-1-3(2-4)5(7)8/h3-4H,1-2H2,(H,7,8)

Inchi Key

AZXXMCROALAIRS-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library