Skip to Content

COM448635566

Combi-Blocks, Inc.

2,3-Dichloro-5-ethoxypyridine

Synonym(s):

2,3-dichloro-5-ethoxypyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
192.04
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H7Cl2NO/c1-2-11-5-3-6(8)7(9)10-4-5/h3-4H,2H2,1H3

Inchi Key

XZBMJARTHCYJRH-UHFFFAOYSA-N

Purity

98%

Country of Origin

CN



Still not finding the right product?

Explore all of our products under 2,3-Dichloro-5-ethoxypyridine


MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?PY-1941



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library