Skip to Content
Merck

3100-OP

CHES

OmniPur® Grade, >= 99.5%

Synonym(s):

OmniPur® CHES, 2-(N-Cyclohexylamino)Ethanesulfonic Acid

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C8H17NO3S
CAS Number:
Molecular Weight:
207.29
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

CHES, OmniPur® Grade, >= 99.5%

InChI

1S/C8H17NO3S.Na/c10-13(11,12)7-6-9-8-4-2-1-3-5-8;/h8-9H,1-7H2,(H,10,11,12);/q;+1/p-1

InChI key

TXWKJOVDGFRVOG-UHFFFAOYSA-M

form

crystals

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

foreign activity

DNAse, none detected
Protease, none detected
RNAse, none detected

shipped in

ambient

storage temp.

10-30°C

Quality Level

General description

Molecular biology grade.

Specifications:
A (260 nm, 50%, water): ≤0.1AU
Assay: ≥99.5%
DNase: None detected
Lead (Pb): ≤0.0001%
Lithium (Li): ≤0.005%
Magnesium (Mg): ≤0.0001%
Nickel (Ni): ≤0.0001%
Potassium (K): ≤0.05%
Protease: None detected
RNase: None detected
Rubidium (Rb): ≤0.1%
Sodium (Na): ≤0.1%





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service