Skip to Content
Merck

STS0026

TERGITOL 15-S-40 solution

70% in water

Synonym(s):

Secondary alcohol ethoxylate 41 EO

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.21

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

laboratory grade

Quality Level

description

non-ionic

form

liquid

mol wt

1,960 g/mol by calculation

concentration

70% in water

CMC

1314 ppm (25°C)

transition temp

cloud point >100 °C (1 wt% actives aq solution)
pour point 5 °C

solubility

water: freely soluble (some compositions may form gels)

density

1.072 g/mL at 30 °C

HLB

18

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is a biodegradable surfactant and is aligned with the 10th principle of Green Chemistry "Design for Degradation".

This product is a 70% aqueous solution of a secondary alcohol ethoxylate with 41 ethylene oxide (EO) units.

Features and Benefits

  • Emulsion stabilizer
  • Provides freeze/thaw & ionic stability
  • Electrolyte solubility
  • Good handling properties

Legal Information

TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service