Skip to Content
Merck

V800442

Urea

≥99%, Vetec, LR grade

Synonym(s):

Carbamide, Carbonyldiamide

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

Linear Formula:
NH2CONH2
CAS Number:
Molecular Weight:
60.06
Beilstein:
635724
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Urea, LR, ≥99%

grade

LR

product line

Vetec

Assay

≥99%

mp

132-135 °C (lit.)

density

1.335 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

NC(N)=O

InChI

1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)

InChI key

XSQUKJJJFZCRTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
V900119RDD03451456
assay

≥99%

assay

99%

assay

≥98%

assay

99% (T)

mp

132-135 °C (lit.)

mp

132-135 °C (lit.)

mp

132-135 °C (lit.)

mp

132-135 °C (lit.)

density

1.335 g/mL at 25 °C (lit.)

density

1.335 g/mL at 25 °C (lit.)

density

1.335 g/mL at 25 °C (lit.)

density

1.335 g/mL at 25 °C (lit.)

grade

LR

grade

reagent grade

grade

-

grade

for molecular biology

product line

Vetec

product line

Vetec

product line

Redi-Dri

product line

BioUltra

functional group

amine

functional group

amine

functional group

-

functional group

-

Application

Used for the denaturation of proteins and as a mild solubilization agent for insoluble or denatured proteins. Useful for renaturing proteins from samples already denatured with 6 M guanidine chloride such as inclusion bodies. May be used with guanidine hydrochloride and dithiothreitrol (DTT) in the refolding of denatured proteins into their native or active form.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Li et al.
Cell death and differentiation, 22(7), 1081-1093 (2014-12-20)
P53 is critically important in preventing oncogenesis but its role in inflammation in general and in the function of inflammatory macrophages in particular is not clear. Here, we show that bone marrow-derived macrophages exhibit endogenous p53 activity, which is increased
Florin Sasarman et al.
Human molecular genetics, 24(10), 2841-2847 (2015-02-06)
Addition of the trinucleotide cytosine/cytosine/adenine (CCA) to the 3' end of transfer RNAs (tRNAs) is essential for translation and is catalyzed by the enzyme TRNT1 (tRNA nucleotidyl transferase), which functions in both the cytoplasm and mitochondria. Exome sequencing revealed TRNT1
Sreenivasa C Ramaiahgari et al.
Archives of toxicology, 88(5), 1083-1095 (2014-03-07)
Immortalized hepatocyte cell lines show only a weak resemblance to primary hepatocytes in terms of gene expression and function, limiting their value in predicting drug-induced liver injury (DILI). Furthermore, primary hepatocytes cultured on two-dimensional tissue culture plastic surfaces rapidly dedifferentiate
A mechanically and electrically self-healing supercapacitor.
Hua Wang et al.
Advanced materials (Deerfield Beach, Fla.), 26(22), 3638-3643 (2014-02-21)
G L French et al.
Lancet (London, England), 2(8551), 117-119 (1987-07-18)
Tuberculostearic acid, a structural component of Mycobacterium tuberculosis, was identified by gas chromatography/mass spectrometry with selected ion monitoring in cerebrospinal fluid (CSF) from 13 patients with proven and 8 out of 9 patients with suspected tuberculous meningitis; the negative result

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service