Merck
Assessing Plant Tolerance to Acute Heat Stress.
Bio-protocol (2017-07-20)
Minsoo Kim, Fionn McLoughlin, Eman Basha, Elizabeth Vierling
ABSTRACT
It is well-established that plants are able to acclimate to temperatures above or below the optimal temperature for their growth. Here, we provide protocols for assays that can be used quantitatively or qualitatively to assess the relative ability of plants to acquire tolerance to high temperature stress. The hypocotyl elongation assay described was developed to screen for mutants defective in the acquisition of tolerance to extreme temperature stress, and other assays were developed to further characterize mutant and transgenic plants for heat tolerance of other processes or at other growth stages. Although the protocols provide details for application to Arabidopsis thaliana, the same basic methods can be adopted to assay heat tolerance in other plant species.
Sigma-Aldrich
Murashige and Skoog Basal Salt Mixture (MS), powder, suitable for plant cell culture
Sigma-Aldrich
MES hydrate, ≥99.5% (titration)
Sigma-Aldrich
Triton X-100, Molecular Biology