Skip to Content
Merck

BDPH9BCC8A4E

Bide Pharmatech

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethoxy)phenol

Synonym(s):

3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethoxy)phenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C13H16O4F3B1
CAS Number:
Molecular Weight:
304.07
MDL number:
Product BDPH9BCC8A4E is not currently sold in your country.


Inchi Code

1S/C13H16BF3O4/c1-11(2)12(3,4)21-14(20-11)9-7-8(18)5-6-10(9)19-13(15,16)17/h5-7,18H,1-4H3

Inchi Key

NDTKUSPSACVYMZ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library