Skip to Content
Merck
Cortisone solution 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Cortisone solution

Cortisone solution 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C21H28O5
CAS Number:
Molecular Weight:
360.44
EC Number:

Products

Product Category
Brand
Manufacturer
Physical Form
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1