Merck
Cholecystokinin and thermoregulation--a minireview.
Peptides (2001-07-18)
Z Szelényi
ABSTRACT
Thermoregulatory effects of cholecystokinin (CCK) peptides are reviewed with special emphasis on two types of responses, that is hypothermia or hyperthermia. In rodents exposed to cold a dose-dependent hypothermia has been observed on peripheral injection of CCK probably acting on CCKA receptors. Central microinjection of CCK in rats induced a thermogenic response that could be attenuated by CCKB receptor antagonists, but some authors observed a hypothermia. It is suggested that neuronal CCK may have a specific role in the development of hyperthermia, and endogenous CCK-ergic mechanisms could contribute to the mediation of fever. Possible connections between thermoregulatory and other autonomic functional changes induced by CCK are discussed.