Merck
  • Exploitation of the broad specificity of the membrane-bound isoenzyme of lactate dehydrogenase for direct selection of null mutants in Neisseria gonorrhoeae.
Exploitation of the broad specificity of the membrane-bound isoenzyme of lactate dehydrogenase for direct selection of null mutants in Neisseria gonorrhoeae.
Journal of general microbiology (1990-01-01)
A T Hendry, R K Bhatnagar, K T Shanmugam, R A Jensen
ABSTRACT
Lactic acid is readily utilized as a carbon and energy source by Neisseria gonorrhoeae. The oxidation of lactate is coupled to electron transport via a membrane-bound lactate dehydrogenase (iLDH) which is independent of pyridine nucleotide. The broad substrate specificity of iLDH endows N. gonorrhoeae with the novel ability to convert phenyllactate to L-phenylalanine via phenylpyruvate. N. gonorrhoeae ATCC 27628 typifies a class of clinical isolate whose growth is inhibited by phenylpyruvate (or L-phenylalanine). Exploiting resistance to growth inhibition by phenyllactate as a strategy of positive selection, mutant derivatives of strain ATCC 27628 lacking iLDH activity were readily obtained. These mutants are incapable of oxidizing phenyllactate, and lack the parent-strain ability to reduce c-type cytochromes in the presence of lactate, phenyllactate or 4-hydroxyphenyllactate. They retain, however, a cytoplasmic NAD(+)-linked lactate dehydrogenase (nLDH). Since the mutants retained the ability to grow on lactate as a sole source of carbon, nLDH presumably can function in an opposite-to-normal physiological direction in the absence of iLDH. This would explain the failure to isolate iLDH-deficient mutants by selection for inability to grow on lactate.
Sigma-Aldrich
DL-p-Hydroxyphenyllactic acid, ≥97% (HPLC)