Merck
Temazepam (Restoril, Sandoz Pharmaceuticals).
Drug intelligence & clinical pharmacy (1982-09-01)
J C McElnay, M E Jones, B Alexander
ABSTRACT
Temazepam is a benzodiazepine derivative indicated for the treatment of insomnia. Pharmacokinetic studies of the hard capsule formulation indicate that the mean time to peak is 2.99 hours and the mean elimination half-life is 14.7 hours. Sleep laboratory studies have demonstrated improvements in all sleep parameters except sleep onset latency. Clinically, patients report improvements in all sleep parameters including sleep onset latency. The efficacy of temazepam compares favorably with barbiturates, glutethimide, nitrazepam, lorazepam, oxazepam, and flurazepam. It has not been compared with diazepam in the clinical setting. Side effects include drowsiness, dizziness, and lethargy. The incidence of hangover effects from 15- and 30-mg doses is relatively low. Temazepam has no proven advantages over other benzodiazepine hypnotics. The major issues that need further clarification include temazepam's sleep induction properties and the relative incidence of hangover and rebound insomnia when compared with longer-acting benzodiazepines.