Merck
Dengue in adults admitted to a referral hospital in Hanoi, Vietnam.
The American journal of tropical medicine and hygiene (2015-04-29)
Walter R Taylor, Annette Fox, Khuong Thi Pham, Hoa Nguyen Minh Le, Ninh Thi Hai Tran, Giang Van Tran, Binh Thanh Nguyen, My Van Nguyen, Lien Thi Nguyen, Sophie Yacoub, Hoai Thi Thu Nguyen, Ha Hong Nguyen, Hien Duc Nguyen, Heiman Wertheim, Peter Horby, Jeremy Farrar, Lien Thi Minh Trinh, Kinh Van Nguyen
ABSTRACT
Knowledge of adult dengue virus (DENV) infection from Hanoi, Vietnam, is limited. In 2008, we prospectively studied 143 (77 male) confirmed (nonstructural 1 antigen enzyme-linked immunosorbent assay [ELISA], DENV polymerase chain reaction, paired serology) adult dengue patients of median age 23.5 (range 16-72) years. They were admitted to the National Hospital for Tropical Diseases, Hanoi, on median illness day (D) 5 (range 1-8). By D8, 141 (98.6%) were afebrile. Platelet counts and hematocrit (median, interquartile range [IQR]) nadired and peaked on D5 and D4, respectively: 40,000/μL (10,000-109,000/μL), 43.4% (34.9-49.7%). Four (2.8%) patients had severe dengue: 1) D10 shock (N = 1) and 2) aspartate aminotransferase (AST) ≥ 1,000 IU/L (N = 3, D5 and D7). Of 143 patients, 118 (82.5%) had ≥ 1 warning sign (World Health Organization [WHO] 2009 criteria): mucosal bleeding 66/143 (46.1%), soft tissue edema 54/143 (37.7%), and ultrasound detected plasma leakage (pleural effusions/ascites) 30/129 (23.25%). 138 (96.5%) patients received intravenous (IV) fluids: 3 L (IQR: 0.5-8.5 L). Most patients had non-severe dengue with warning signs. High rates of edema and plasma leakage may be explained partly by liberal use of IV fluids. Studies are needed on optimizing fluid management in non-severe adult dengue.