Skip to Content
Merck

BDPH9BCD6058

Bide Pharmatech

9,10-Dimethoxy-5,6-dihydro-[1,3]dioxolo[4,5-g]isoquinolino[3,2-a]isoquinolin-7-ium chloride

Synonym(s):

9,10-dimethoxy-5H,6H-[1,3]dioxolo[4,5-g]isoquino[3,2-a]isoquinolin-7-ium chloride

Select a Size

Change View

About This Item

Linear Formula:
C20H18O4N1Cl1
CAS Number:
Molecular Weight:
371.82
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C20H18NO4.ClH/c1-22-17-4-3-12-7-16-14-9-19-18(24-11-25-19)8-13(14)5-6-21(16)10-15(12)20(17)23-2;/h3-4,7-10H,5-6,11H2,1-2H3;1H/q+1;/p-1

Inchi Key

VKJGBAJNNALVAV-UHFFFAOYSA-M

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Form: solid




Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library