Skip to Content
Merck

BDPH9C01EDC2

Bide Pharmatech

(2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol

Synonym(s):

1-deoxy-1-(methylamino)-D-glucitol

Select a Size

Change View

About This Item

Linear Formula:
C7H17O5N1
CAS Number:
Molecular Weight:
195.22
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H17NO5/c1-8-2-4(10)6(12)7(13)5(11)3-9/h4-13H,2-3H2,1H3/t4-,5+,6+,7+/m0/s1

Inchi Key

MBBZMMPHUWSWHV-BDVNFPICSA-N

Shipping Temperature

Normal

Purity

99%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Form: Crystal - Powder / Colour: White - Almost white

Melting Point

129-131.5C



Still not finding the right product?


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library