Skip to Content
Merck

CIAH987EC4A4

ChemScene LLC (Preferred Partner)

N,N'-(R)-[1,1'-Binaphthalene]-2,2'-diylbis[N'-[3,5-bis(trifluoromethyl)phenyl]thiourea]

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
826.73
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

bp

676.4+-65.0 C at 760 mmHg

mp

132-134 C

Iupac Name

1,1'-([1,1'-binaphthalene]-2,2'-diyl)bis(3-(3,5-bis(trifluoromethyl)phenyl)thiourea)

Inchi Code

1S/C38H22F12N4S2/c39-35(40,41)21-13-22(36(42,43)44)16-25(15-21)51-33(55)53-29-11-9-19-5-1-3-7-27(19)31(29)32-28-8-4-2-6-20(28)10-12-30(32)54-34(56)52-26-17-23(37(45,46)47)14-24(18-26)38(48,49)50/h1-18H,(H2,51,53,55)(H2,52,54,56)

InChI key

OMOQSDHNLKLWHK-UHFFFAOYSA-N

storage temp.

4C, protect from light, stored under nitrogen

Purity

95%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/1005003-43-9.html

Boiling point

676.4+-65.0 C at 760 mmHg

Melting Point

132-134 C

Signal Word

Warning

Hazard Statements

Precautionary statements


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service