Skip to Content
Merck
Blood typing Intermediates for Further Manufacturing Use (FFMU) - RHCE (Rh) Specificities Anti-C from the clone MS-273, Human IgM, Intermediate for further manufacturing use

Blood typing Intermediates for Further Manufacturing Use (FFMU) - RHCE (Rh) Specificities

Blood typing Intermediates for Further Manufacturing Use (FFMU) - RHCE (Rh) Specificities Anti-C from the clone MS-273, Human IgM, Intermediate for further manufacturing use

Products

Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Quality Segment
Package Size
Technique
Showing 1-11 of 11
Compare
Product Number
Description
Pricing