Merck
  • Ectopic expression of the Brassica SHOOTMERISTEMLESS attenuates the deleterious effects of the auxin transport inhibitor TIBA on somatic embryo number and morphology.
Ectopic expression of the Brassica SHOOTMERISTEMLESS attenuates the deleterious effects of the auxin transport inhibitor TIBA on somatic embryo number and morphology.
Plant science : an international journal of experimental plant biology (2011-03-23)
Mohamed Elhiti, Claudio Stasolla
ABSTRACT
The auxin transport inhibitor 2,3,5-triiodobenzoic acid (TIBA) is a useful compound for investigating the role of auxin flow during plant growth and development. In Arabidopsis lines, applications of TIBA during the induction phase of somatic embryogenesis inhibit embryo development and induce the differentiation of the meristematic cells of the shoot apical meristem (SAM), leading to the fusion of the cotyledons. These abnormalities were associated to changes in the expression levels of auxin transporter genes (PINs) and endogenous distribution of IAA. Treatments of TIBA caused a rapid accumulation of IAA within the epidermal and cortical root cells of the explants (bent-cotyledon zygotic embryos), as well as in the apical and sub-apical cells of the callus generated by the surface of the cotyledons of the explants. Within the callus only a few cells acquired meristematic characteristics, and this was associated to low expression levels of genes involved in embryogenic cell fate acquisition, such as WUSCHEL (WUS), LEAFY COTYLEDON 1 and 2. All these deleterious effects were attenuated when TIBA was administered to lines over-expressing SHOOT MERISTEMLESS (STM) isolated from Brassica oleracea (Bo), B. napus (Bn), and B. rapa (Br). Of interest, TIBA-treated explants of Arabidopsis lines over-expressing the Brassica STM were able to produce a large number of embryogenic cells and somatic embryos which exhibited a normal morphology and two distinct cotyledons. A proposed reason for this behaviour was ascribed to the ability of the transformed tissue to retain a normal distribution of auxin in the presence of TIBA. Proper localization of auxin might be required for the normal expression of several genes needed for the acquisition of embryogenic competence and formation of somatic embryos.
Sigma-Aldrich
2,3,5-Triiodobenzoic acid, BioReagent, suitable for plant cell culture, ≥97% (HPLC)