Merck
  • Jasmonic acid distribution and action in plants: regulation during development and response to biotic and abiotic stress.
Jasmonic acid distribution and action in plants: regulation during development and response to biotic and abiotic stress.
Proceedings of the National Academy of Sciences of the USA (1995)
Creelman AR and Mullet EJ