Merck
Proteogenomic Analysis of Surgically Resected Lung Adenocarcinoma.
Journal of thoracic oncology : official publication of the International Association for the Study of Lung Cancer (2018-07-19)
Michael F Sharpnack, Nilini Ranbaduge, Arunima Srivastava, Ferdinando Cerciello, Simona G Codreanu, Daniel C Liebler, Celine Mascaux, Wayne O Miles, Robert Morris, Jason E McDermott, James L Sharpnack, Joseph Amann, Christopher A Maher, Raghu Machiraju, Vicki H Wysocki, Ramaswami Govindan, Parag Mallick, Kevin R Coombes, Kun Huang, David P Carbone
ABSTRACT
Despite apparently complete surgical resection, approximately half of resected early-stage lung cancer patients relapse and die of their disease. Adjuvant chemotherapy reduces this risk by only 5% to 8%. Thus, there is a need for better identifying who benefits from adjuvant therapy, the drivers of relapse, and novel targets in this setting. RNA sequencing and liquid chromatography/liquid chromatography-mass spectrometry proteomics data were generated from 51 surgically resected non-small cell lung tumors with known recurrence status. We present a rationale and framework for the incorporation of high-content RNA and protein measurements into integrative biomarkers and show the potential of this approach for predicting risk of recurrence in a group of lung adenocarcinomas. In addition, we characterize the relationship between mRNA and protein measurements in lung adenocarcinoma and show that it is outcome specific. Our results suggest that mRNA and protein data possess independent biological and clinical importance, which can be leveraged to create higher-powered expression biomarkers.
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95% (titration)