Merck
  • Multiplex quantitative assays indicate a need for reevaluating reported small-molecule TrkB agonists.
Multiplex quantitative assays indicate a need for reevaluating reported small-molecule TrkB agonists.
Science signaling (2017-08-24)
Umed Boltaev, Yves Meyer, Farangis Tolibzoda, Teresa Jacques, Madalee Gassaway, Qihong Xu, Florence Wagner, Yan-Ling Zhang, Michelle Palmer, Edward Holson, Dalibor Sames
ABSTRACT
Brain-derived neurotrophic factor (BDNF) and its receptor, tropomyosin-related kinase B (TrkB), have emerged as key regulators of brain plasticity and represent disease-modifying targets for several brain disorders, including Alzheimer's disease and major depressive disorder. Because of poor pharmacokinetic properties of BDNF, the interest in small-molecule TrkB agonists and modulators is high. Several compounds have been reported to act as TrkB agonists, and their increasing use in various nervous system disorder models creates the perception that these are reliable probes. To examine key pharmacological parameters of these compounds in detail, we have developed and optimized a series of complementary quantitative assays that measure TrkB receptor activation, TrkB-dependent downstream signaling, and gene expression in different cellular contexts. Although BDNF and other neurotrophic factors elicited robust and dose-dependent receptor activation and downstream signaling, we were unable to reproduce these activities using the reported small-molecule TrkB agonists. Our findings indicate that experimental results obtained with these compounds must be carefully interpreted and highlight the challenge of developing reliable pharmacological activators of this key molecular target.
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Anti-TrkB antibody produced in goat, affinity isolated antibody, lyophilized powder