Merck
  • Iron-catalyzed selective biaryl coupling: Remarkable suppression of homocoupling by the fluoride anion.
Iron-catalyzed selective biaryl coupling: Remarkable suppression of homocoupling by the fluoride anion.
Journal of the American Chemical Society (2007)
Hatakeyama T and Nakamura M