Merck
  • A myocardial slice culture model reveals alpha-1A-adrenergic receptor signaling in the human heart
A myocardial slice culture model reveals alpha-1A-adrenergic receptor signaling in the human heart
JACC. Basic to translational science (2016)
Thomas RC, et al.