- Bacterial chemoattraction towards jasmonate plays a role in the entry of Dickeya dadantii through wounded tissues
Bacterial chemoattraction towards jasmonate plays a role in the entry of Dickeya dadantii through wounded tissues
Molecular Microbiology (2009)
Antunez-Lamas M, et al.