Merck
  • Novel insights into catalytic mechanism from a crystal structure of human topoisomerase I in complex with DNA
Novel insights into catalytic mechanism from a crystal structure of human topoisomerase I in complex with DNA
Biochemistry (2000)
Redinbo MR, et al.