Merck
  • Protein crystallization in agarose gel with high strength: developing an automated system for protein crystallographic processes
Protein crystallization in agarose gel with high strength: developing an automated system for protein crystallographic processes
Japanese Journal of Applied Physics (2009)
Sugiyama S, et al.