- Foundations of identifying individual chromosomes by imaging flow cytometry with applications in radiation biodosimetry
Foundations of identifying individual chromosomes by imaging flow cytometry with applications in radiation biodosimetry
Methods (2017)
Beaton-Green LA, et al.