Merck
  • Identification and quantification of para-phenylenediamine in a temporary black henna tattoo
Identification and quantification of para-phenylenediamine in a temporary black henna tattoo
American Journal of Contact Dermatitis : Official Journal of the American Contact Dermatitis Society (2002)
Brancaccio RR, et al.