Merck
Fcμ receptor as a Costimulatory Molecule for T Cells.
Cell reports (2019-03-07)
Andreas Meryk, Luca Pangrazzi, Magdalena Hagen, Florian Hatzmann, Brigitte Jenewein, Bojana Jakic, Natascha Hermann-Kleiter, Gottfried Baier, Juulia Jylhävä, Mikko Hurme, Klemens Trieb, Beatrix Grubeck-Loebenstein
ABSTRACT
Fc receptor for IgM (FcμR)-deficient mice display dysregulated function of neutrophils, dendritic cells, and B cells. The relevance of FcμR to human T cells is still unknown. We show that FcμR is mostly stored inside the cell and that surface expression is tightly regulated. Decreased surface expression on T cells from elderly individuals is associated with alterations in the methylation pattern of the FCMR gene. Binding and internalization of IgM stimulate transport of FcμR to the cell surface to ensure sustained IgM uptake. Concurrently, IgM accumulates within the cell, and the surface expression of other receptors increases, among them the T cell receptor (TCR) and costimulatory molecules. This leads to enhanced TCR signaling, proliferation, and cytokine release, in response to low, but not high, doses of antigen. Our findings indicate that FcμR is an important regulator of T cell function and reveal an additional mode of interaction between B and T cells.
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, Molecular Biology
Sigma-Aldrich
Anti-FCMR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
IgM from human serum, reagent grade, ~95% (HPLC), buffered aqueous solution
Sigma-Aldrich
MG-132, ≥95% by HPLC, Potent, reversible, and cell-permeable proteasome inhibitor (Ki = 4 nM).