Merck
  • Multiple antibiotic resistance indexing of Escherichia coli to identify high-risk sources of fecal contamination of foods.
Multiple antibiotic resistance indexing of Escherichia coli to identify high-risk sources of fecal contamination of foods.
Applied and Environmental Microbiology (1983)
Krumperman PH, et al.