Merck
  • Antimicrobial Photodynamic Therapy (APDT) Action Based on Nanostructured Photosensitizers
Antimicrobial Photodynamic Therapy (APDT) Action Based on Nanostructured Photosensitizers
Multifunctional Systems for Combined Delivery Biosensing and Diagnostics (2017)