Merck
  • A new HPLC method for simultaneously measuring chloride, sugars, organic acids and alcohols in food samples
A new HPLC method for simultaneously measuring chloride, sugars, organic acids and alcohols in food samples
J. Food Compos. Anal. (2017)
Zaky AS