Merck
  • Simultaneous Determination of Succinic Acid and Water-Soluble Vitamins by Ion-Pair High-Performance Liquid Chromatography
Simultaneous Determination of Succinic Acid and Water-Soluble Vitamins by Ion-Pair High-Performance Liquid Chromatography
Journal of Analytical Chemistry (2018)
Leonov KA