Merck
  • A stromal cell niche sustains ILC2-mediated type-2 conditioning in adipose tissue.
A stromal cell niche sustains ILC2-mediated type-2 conditioning in adipose tissue.
The Journal of experimental medicine (2019-06-30)
Batika M J Rana, Eric Jou, Jillian L Barlow, Noe Rodriguez-Rodriguez, Jennifer A Walker, Claire Knox, Helen E Jolin, Clare S Hardman, Meera Sivasubramaniam, Aydan Szeto, E Suzanne Cohen, Ian C Scott, Matthew A Sleeman, Chiamaka I Chidomere, Sara Cruz Migoni, Jorge Caamano, Helle F Jorgensen, Stefania Carobbio, Antonio Vidal-Puig, Andrew N J McKenzie
ABSTRACT
Group-2 innate lymphoid cells (ILC2), type-2 cytokines, and eosinophils have all been implicated in sustaining adipose tissue homeostasis. However, the interplay between the stroma and adipose-resident immune cells is less well understood. We identify that white adipose tissue-resident multipotent stromal cells (WAT-MSCs) can act as a reservoir for IL-33, especially after cell stress, but also provide additional signals for sustaining ILC2. Indeed, we demonstrate that WAT-MSCs also support ICAM-1-mediated proliferation and activation of LFA-1-expressing ILC2s. Consequently, ILC2-derived IL-4 and IL-13 feed back to induce eotaxin secretion from WAT-MSCs, supporting eosinophil recruitment. Thus, MSCs provide a niche for multifaceted dialogue with ILC2 to sustain a type-2 immune environment in WAT.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Geranyl pyrophosphate ammonium salt, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)