Merck
  • Stearic acid induces proinflammatory cytokine production partly through activation of lactate-HIF1α pathway in chondrocytes.
Stearic acid induces proinflammatory cytokine production partly through activation of lactate-HIF1α pathway in chondrocytes.
Scientific reports (2015-08-15)
Hongming Miao, Liang Chen, Lijun Hao, Xuan Zhang, Yujuan Chen, Zhihua Ruan, Houjie Liang
ABSTRACT
The biomechanics stress and chronic inflammation in obesity are causally linked to osteoarthritis. However, the metabolic factors mediating obesity-related osteoarthritis are still obscure. Here we scanned and identified at least two elevated metabolites (stearic acid and lactate) from the plasma of diet-induced obese mice. We found that stearic acid potentiated LDH-a-dependent production of lactate, which further stabilized HIF1α protein and increased VEGF and proinflammatory cytokine expression in primary mouse chondrocytes. Treatment with LDH-a and HIF1α inhibitors notably attenuated stearic acid-or high fat diet-stimulated proinflammatory cytokine production in vitro and in vivo. Furthermore, positive correlation of plasma lactate, cartilage HIF1α and cytokine levels with the body mass index was observed in subjects with osteoarthritis. In conclusion, saturated free fatty acid induced proinflammatory cytokine production partly through activation of a novel lactate-HIF1α pathway in chondrocytes. Our findings hold promise of developing novel clinical strategies for the management of obesity-related diseases such as osteoarthritis.
Sigma-Aldrich
TLR4-IN-C34, ≥98% (HPLC)
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)