Merck
  • Origins of Neural Progenitor Cell-Derived Axons Projecting Caudally after Spinal Cord Injury.
Origins of Neural Progenitor Cell-Derived Axons Projecting Caudally after Spinal Cord Injury.
Stem cell reports (2019-06-18)
Paul Lu, Walace Gomes-Leal, Selin Anil, Gabriel Dobkins, J Russell Huie, Adam R Ferguson, Lori Graham, Mark Tuszynski
ABSTRACT
Neural progenitor cells (NPCs) transplanted into sites of spinal cord injury (SCI) extend large numbers of axons into the caudal host spinal cord. We determined the precise locations of neurons in the graft that extend axons into the caudal host spinal cord using AAV9-Cre-initiated retrograde tracing into floxed-TdTomato-expressing NPC grafts. 7,640 ± 630 grafted neurons extended axons to a single caudal host spinal cord site located 2 mm beyond the lesion, 5 weeks post injury. While caudally projecting axons arose from neurons located in all regions of the graft, the majority of caudally projecting graft neurons (53%) were located within the caudal one-third of the graft. Numerous host corticospinal axons formed monosynaptic projections onto caudally projecting graft neurons; however, we find that the majority of host axonal neuronal projections formed by neural progenitor cell interneuronal "relays" across sites of SCI are likely polysynaptic in nature.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-MAP2A, 2B, 2C Antibody, clone HM-2, ascites fluid, clone HM-2, Chemicon®
Sigma-Aldrich
Anti-Synaptophysin Antibody, clone SY38, clone SY38, from mouse