Merck
  • Clinical Characteristics of 58 Children With a Pediatric Inflammatory Multisystem Syndrome Temporally Associated With SARS-CoV-2.
Clinical Characteristics of 58 Children With a Pediatric Inflammatory Multisystem Syndrome Temporally Associated With SARS-CoV-2.
JAMA (2020-06-09)
Elizabeth Whittaker, Alasdair Bamford, Julia Kenny, Myrsini Kaforou, Christine E Jones, Priyen Shah, Padmanabhan Ramnarayan, Alain Fraisse, Owen Miller, Patrick Davies, Filip Kucera, Joe Brierley, Marilyn McDougall, Michael Carter, Adriana Tremoulet, Chisato Shimizu, Jethro Herberg, Jane C Burns, Hermione Lyall, Michael Levin
ABSTRACT
In communities with high rates of coronavirus disease 2019, reports have emerged of children with an unusual syndrome of fever and inflammation. To describe the clinical and laboratory characteristics of hospitalized children who met criteria for the pediatric inflammatory multisystem syndrome temporally associated with severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) (PIMS-TS) and compare these characteristics with other pediatric inflammatory disorders. Case series of 58 children from 8 hospitals in England admitted between March 23 and May 16, 2020, with persistent fever and laboratory evidence of inflammation meeting published definitions for PIMS-TS. The final date of follow-up was May 22, 2020. Clinical and laboratory characteristics were abstracted by medical record review, and were compared with clinical characteristics of patients with Kawasaki disease (KD) (n = 1132), KD shock syndrome (n = 45), and toxic shock syndrome (n = 37) who had been admitted to hospitals in Europe and the US from 2002 to 2019. Signs and symptoms and laboratory and imaging findings of children who met definitional criteria for PIMS-TS from the UK, the US, and World Health Organization. Clinical, laboratory, and imaging characteristics of children meeting definitional criteria for PIMS-TS, and comparison with the characteristics of other pediatric inflammatory disorders. Fifty-eight children (median age, 9 years [interquartile range {IQR}, 5.7-14]; 20 girls [34%]) were identified who met the criteria for PIMS-TS. Results from SARS-CoV-2 polymerase chain reaction tests were positive in 15 of 58 patients (26%) and SARS-CoV-2 IgG test results were positive in 40 of 46 (87%). In total, 45 of 58 patients (78%) had evidence of current or prior SARS-CoV-2 infection. All children presented with fever and nonspecific symptoms, including vomiting (26/58 [45%]), abdominal pain (31/58 [53%]), and diarrhea (30/58 [52%]). Rash was present in 30 of 58 (52%), and conjunctival injection in 26 of 58 (45%) cases. Laboratory evaluation was consistent with marked inflammation, for example, C-reactive protein (229 mg/L [IQR, 156-338], assessed in 58 of 58) and ferritin (610 μg/L [IQR, 359-1280], assessed in 53 of 58). Of the 58 children, 29 developed shock (with biochemical evidence of myocardial dysfunction) and required inotropic support and fluid resuscitation (including 23/29 [79%] who received mechanical ventilation); 13 met the American Heart Association definition of KD, and 23 had fever and inflammation without features of shock or KD. Eight patients (14%) developed coronary artery dilatation or aneurysm. Comparison of PIMS-TS with KD and with KD shock syndrome showed differences in clinical and laboratory features, including older age (median age, 9 years [IQR, 5.7-14] vs 2.7 years [IQR, 1.4-4.7] and 3.8 years [IQR, 0.2-18], respectively), and greater elevation of inflammatory markers such as C-reactive protein (median, 229 mg/L [IQR 156-338] vs 67 mg/L [IQR, 40-150 mg/L] and 193 mg/L [IQR, 83-237], respectively). In this case series of hospitalized children who met criteria for PIMS-TS, there was a wide spectrum of presenting signs and symptoms and disease severity, ranging from fever and inflammation to myocardial injury, shock, and development of coronary artery aneurysms. The comparison with patients with KD and KD shock syndrome provides insights into this syndrome, and suggests this disorder differs from other pediatric inflammatory entities.