Merck
  • Enhancement of coffee brew aroma through control of the aroma staling pathway of 2-furfurylthiol.
Enhancement of coffee brew aroma through control of the aroma staling pathway of 2-furfurylthiol.
Food chemistry (2020-04-14)
Zhenchun Sun, Heping Cui, Ni Yang, Charfedinne Ayed, Xiaoming Zhang, Ian D Fisk
ABSTRACT
During storage of coffee, the key aroma 2-furfurylthiol becomes less active, the mechanisms of this loss and ways to mitigate it were investigated. Aroma profiles were analyzed using GC-MS and sensory properties were evaluated by Quantitative Descriptive Analysis. Quinones, as the oxidation products of hydroxydroquinone, was found to actively bind 2-furfurylthiol, which accounted for the loss of 2-furfurylthiol. To mitigate this loss, ingredients were screened for their ability to prevent 2-furfurylthiol from loss. Cysteine had the highest 2-furfurylthiol releasing efficiency and ascorbic acid was also selected due to its 2-furfurylthiol releasing ability in Fenton reaction system. Concentrations were optimized and the addition of 0.045 g/L cysteine and 0.05 g/L ascorbic acid directly protected aroma during storage, these included 2-furfurylthiol, dimethyltrisulfide, methyl furfuryl disulfide, 4-ethylguaiacol and 4-vinylguaiacol. Ultimately, sensory testing showed a direct enhancement in nutty, sulfurous and roasted aroma attributes, an increase in flavour intensity and preference over shelf life.
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
1,2,4-Benzenetriol, ReagentPlus®, 99%