Merck
  • Phytochemistry and antifungal properties of the newly discovered tree Pleodendron costaricense.
Phytochemistry and antifungal properties of the newly discovered tree Pleodendron costaricense.
Journal of natural products (2006-07-29)
Virginie Treyvaud Amiguet, Philippe Petit, Chieu Anh Ta, Ronaldo Nuñez, Pablo Sánchez-Vindas, Luis Poveda Alvarez, Myron L Smith, John Thor Arnason, Tony Durst
ABSTRACT
Gas chromatography analysis of the essential oils of leaves and bark collected from the newly discovered tree Pleodendron costaricense identified alpha-pinene, beta-pinene, beta-myrcene, beta-thujene, and beta-caryophyllene as their major constituents. Phytochemical analysis of P. costaricense parts led to the isolation and identification of delta-tocotrienol, beta-sitosterol, four known drimane-type sesquiterpenes, cinnamodial (1), cinnamosmolide (2), polygodial (3), and mukaadial (4), and two new compounds, a drimane-type sesquiterpene, parritadial (5), and an eremophilane-type sesquiterpene, pleodendione (6). Antifungal assays with the two major compounds, 1 and 2, were carried out, and results showed a high activity of 1 against Alternaria alternata (MIC = 3.9 microg/mL), Candida albicans azole-resistant strain D10, and Wangiella dermatitides (MICs = 15.6 microg/mL). Compound 2 showed less potent antifungal activities than 1 but was more effective against Candida albicans azole-resistant strain CN1A (MIC = 23.4 microg/mL) and Pseudallescheria boydii (MIC = 78.1 microg/mL). A combination of the dialdehyde sesquiterpenes with dillapiol showed a synergistic antifungal effect with 1 and an additive effect with 4 and 5.
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
Sigma-Aldrich
Berberine chloride form