Merck
MoS2/graphene composite paper for sodium-ion battery electrodes.
ACS nano (2014-01-23)
Lamuel David, Romil Bhandavat, Gurpreet Singh
ABSTRACT
We study the synthesis and electrochemical and mechanical performance of layered free-standing papers composed of acid-exfoliated few-layer molybdenum disulfide (MoS2) and reduced graphene oxide (rGO) flakes for use as a self-standing flexible electrode in sodium-ion batteries. Synthesis was achieved through vacuum filtration of homogeneous dispersions consisting of varying weight percent of acid-treated MoS2 flakes in GO in DI water, followed by thermal reduction at elevated temperatures. The electrochemical performance of the crumpled composite paper (at 4 mg cm(-2)) was evaluated as counter electrode against pure Na foil in a half-cell configuration. The electrode showed good Na cycling ability with a stable charge capacity of approximately 230 mAh g(-1) with respect to total weight of the electrode with Coulombic efficiency reaching approximately 99%. In addition, static uniaxial tensile tests performed on crumpled composite papers showed high average strain to failure reaching approximately 2%.
Sigma-Aldrich
Graphene oxide, organic solvent dispersible