Merck
  • A simple method of hiPSCs differentiation into insulin-producing cells is improved with vitamin C and RepSox.
A simple method of hiPSCs differentiation into insulin-producing cells is improved with vitamin C and RepSox.
PloS one (2021-07-13)
Ayumi Horikawa, Keiko Mizuno, Kyoko Tsuda, Takayoshi Yamamoto, Tatsuo Michiue
ABSTRACT
Human induced pluripotent stem cells (hiPSCs) are considered a promising source of pancreatic β-cells for the treatment of diabetes. However, this approach is limited by issues such as low efficiency and high cost. Here, we have developed a new protocol to induce insulin-producing cells. To reduce costs, we decreased the number of reagents and replaced protein reagents with chemical compounds. In this method, we increased induction efficiency with ascorbic acid (vitamin C) and an ALK5 inhibitor, RepSox. In 2D culture, the majority of cells were immature β-cells with low glucose-stimulated insulin secretion. Transferring to 3D culture immediately after endocrine progenitor cell differentiation, however, improved glucose-stimulated insulin secretion. This simplified method will contribute to realizing transplantation therapy of β-cells using iPSCs.
Sigma-Aldrich
Monoclonal Anti-Insulin antibody produced in mouse, clone K36AC10, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Glucagon antibody produced in mouse, clone K79bB10, ascites fluid