Merck
  • Lipid-gated monovalent ion fluxes regulate endocytic traffic and support immune surveillance.
Lipid-gated monovalent ion fluxes regulate endocytic traffic and support immune surveillance.
Science (New York, N.Y.) (2019-12-07)
Spencer A Freeman, Stefan Uderhardt, Amra Saric, Richard F Collins, Catherine M Buckley, Sivakami Mylvaganam, Parastoo Boroumand, Jonathan Plumb, Ronald N Germain, Dejian Ren, Sergio Grinstein
ABSTRACT
Despite ongoing (macro)pinocytosis of extracellular fluid, the volume of the endocytic pathway remains unchanged. To investigate the underlying mechanism, we used high-resolution video imaging to analyze the fate of macropinosomes formed by macrophages in vitro and in situ. Na+, the primary cationic osmolyte internalized, exited endocytic vacuoles via two-pore channels, accompanied by parallel efflux of Cl- and osmotically coupled water. The resulting shrinkage caused crenation of the membrane, which fostered recruitment of curvature-sensing proteins. These proteins stabilized tubules and promoted their elongation, driving vacuolar remodeling, receptor recycling, and resolution of the organelles. Failure to resolve internalized fluid impairs the tissue surveillance activity of resident macrophages. Thus, osmotically driven increases in the surface-to-volume ratio of endomembranes promote traffic between compartments and help to ensure tissue homeostasis.
Sigma-Aldrich
Ouabain octahydrate, ≥95% (HPLC), powder
Sigma-Aldrich
Deoxyribonuclease II from bovine spleen, Type V, essentially salt-free, lyophilized powder, ≥1,000 units/mg protein
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Nocodazole, Inhibitor of mitosis.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
WX8, ≥98% (HPLC)