Merck
  • Increased apoptotic sensitivity of glioblastoma enables therapeutic targeting by BH3-mimetics.
Increased apoptotic sensitivity of glioblastoma enables therapeutic targeting by BH3-mimetics.
Cell death and differentiation (2022-04-28)
Anna L Koessinger, Catherine Cloix, Dominik Koessinger, Dieter Henrik Heiland, Florian J Bock, Karen Strathdee, Kevin Kinch, Laura Martínez-Escardó, Nikki R Paul, Colin Nixon, Gaurav Malviya, Mark R Jackson, Kirsteen J Campbell, Katrina Stevenson, Sandeep Davis, Yassmin Elmasry, Asma Ahmed, Jim O'Prey, Gabriel Ichim, Oliver Schnell, William Stewart, Karen Blyth, Kevin M Ryan, Anthony J Chalmers, Jim C Norman, Stephen W G Tait
ABSTRACT
Glioblastoma (GBM) is the most prevalent malignant primary brain tumour in adults. GBM typically has a poor prognosis, mainly due to a lack of effective treatment options leading to tumour persistence or recurrence. We investigated the therapeutic potential of targeting anti-apoptotic BCL-2 proteins in GBM. Levels of anti-apoptotic BCL-xL and MCL-1 were consistently increased in GBM compared with non-malignant cells and tissue. Moreover, we found that relative to their differentiated counterparts, patient-derived GBM stem-like cells also displayed higher expression of anti-apoptotic BCL-2 family members. High anti-apoptotic BCL-xL and MCL-1 expression correlated with heightened susceptibility of GBM to BCL-2 family protein-targeting BH3-mimetics. This is indicative of increased apoptotic priming. Indeed, GBM displayed an obligate requirement for MCL-1 expression in both tumour development and maintenance. Investigating this apoptotic sensitivity, we found that sequential inhibition of BCL-xL and MCL-1 led to robust anti-tumour responses in vivo, in the absence of overt toxicity. These data demonstrate that BCL-xL and MCL-1 pro-survival function is a fundamental prerequisite for GBM survival that can be therapeutically exploited by BH3-mimetics.
Sigma-Aldrich
Monoclonal Anti-Actin antibody produced in mouse, clone AC-40, ascites fluid
Sigma-Aldrich
Temozolomide, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2