Merck
  • The trophectoderm acts as a niche for the inner cell mass through C/EBPα-regulated IL-6 signaling.
The trophectoderm acts as a niche for the inner cell mass through C/EBPα-regulated IL-6 signaling.
Stem cell reports (2022-08-13)
Marcos Plana-Carmona, Gregoire Stik, Romain Bulteau, Carolina Segura-Morales, Noelia Alcázar, Chris D R Wyatt, Antonios Klonizakis, Luisa de Andrés-Aguayo, Maxime Gasnier, Tian V Tian, Guillem Torcal Garcia, Maria Vila-Casadesús, Nicolas Plachta, Manuel Serrano, Mirko Francesconi, Thomas Graf
ABSTRACT
IL-6 has been shown to be required for somatic cell reprogramming into induced pluripotent stem cells (iPSCs). However, how Il6 expression is regulated and whether it plays a role during embryo development remains unknown. Here, we describe that IL-6 is necessary for C/EBPα-enhanced reprogramming of B cells into iPSCs but not for B cell to macrophage transdifferentiation. C/EBPα overexpression activates both Il6 and Il6ra genes in B cells and in PSCs. In embryo development, Cebpa is enriched in the trophectoderm of blastocysts together with Il6, while Il6ra is mostly expressed in the inner cell mass (ICM). In addition, Il6 expression in blastocysts requires Cebpa. Blastocysts secrete IL-6 and neutralization of the cytokine delays the morula to blastocyst transition. The observed requirement of C/EBPα-regulated IL-6 signaling for pluripotency during somatic cell reprogramming thus recapitulates a physiologic mechanism in which the trophectoderm acts as niche for the ICM through the secretion of IL-6.
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
ESGRO® Recombinant Mouse LIF Protein, ESGRO Leukemia Inhibitory Factor (LIF) supplement for mouse ES cell culture. Each vial contains 10^6 units/ml