Merck
  • MgI2-Catalyzed Nucleophilic Ring-Opening Reactions of Donor-Acceptor Cyclopropanes with Indoline-2-thiones.
MgI2-Catalyzed Nucleophilic Ring-Opening Reactions of Donor-Acceptor Cyclopropanes with Indoline-2-thiones.
The Journal of organic chemistry (2022-08-03)
Pan Tang, You-Yuan Wei, Long Wen, Hao-Jie Ma, Yi Yang, Yan Jiang
ABSTRACT
MgI2-catalyzed nucleophilic ring-opening reactions of donor-acceptor cyclopropanes with indoline-2-thiones as easy-to-handle sulfur nucleophiles were investigated. A series of functionalized γ-indolylthio butyric acid derivatives were synthesized in good to excellent yields under mild reaction conditions. Furthermore, the thioether functionalized ring-opening products could be transformed to sulfone and methionine analogues.
Sigma-Aldrich
Indoline-2-thione, ≥-95%