Merck
  • N-Alkylated Analogues of Indolylthio Glycosides as Glycosyl Donors with Enhanced Activation Profile.
N-Alkylated Analogues of Indolylthio Glycosides as Glycosyl Donors with Enhanced Activation Profile.
European journal of organic chemistry (2022-11-08)
Ganesh Shrestha, Matteo Panza, Yashapal Singh, Keith J Stine, Alexei V Demchenko
ABSTRACT
While studying indolylthio glycosides, previously we determined their activation profile that required large excess of activators. This drawback was partially addressed in the present study of N-alkylated SInR derivatives. The activation process was studied by NMR and the increased understanding of the mechanism led to a discovery of different activation pathways taking place with SIn versus SInR derivatives. Also investigated was orthogonality of the SInR leaving groups versus thioglycosides and selective activation of thioimidates over SInR glycosides.
Sigma-Aldrich
1-Methylindoline-2-thione, ≥95%